Learn More
Cayman Chemical NOLosartan A Other Receptor Mo
SDP
Catalog No. 502890042
Encompass_Preferred
A potent AT1 receptor antagonist linked to an NO-donor; possesses similar anti-hypertensive effects to losartan, with the addition of the vasodilating effects of NO release
Catalog No. 50-289-0042 Supplier Cayman Chemical Supplier No. 1000645610 MG
A potent AT1 receptor antagonist linked to an NO-donor; possesses similar anti-hypertensive effects to losartan, with the addition of the vasodilating effects of NO release