Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nomega-Amino-L-arginine hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH58861MF
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58861MF 50 mg
AAH58861MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH58861MF Supplier Thermo Scientific Chemicals Supplier No. H58861MF

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031799-40-2
Molecular Formula C6H16ClN5O2
Molecular Weight (g/mol) 225.68
MDL Number MFCD00672614
InChI Key YGVMJVSXFJRHGQ-UHFFFAOYNA-N
Synonym nomega-amino-l-arginine hydrochloride, 2s-2-amino-5-n'-aminocarbamimidamido pentanoic acid hydrochloride, ng-amino-l-arginine hydrochloride, c6h15n5o2.hcl, s-2-amino-5-hydrazinecarboximidamido pentanoic acid hydrochloride
PubChem CID 91886146
SMILES Cl.NNC(N)=NCCCC(N)C(O)=O

Specifications

IUPAC Name (2S)-2-amino-5-[[amino(hydrazinyl)methylidene]amino]pentanoic acid;hydrochloride
Quantity 50 mg
Formula Weight 225.67
Percent Purity 97%
Sensitivity Hygroscopic
Chemical Name or Material Nomega-Amino-L-arginine hydrochloride
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.