Learn More
TARGETMOL CHEMICALS INC Nomegestrol acetate 25MG
SDP
Catalog No. 502876990
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Nomegestrol acetate is a widely used progestin-like drug,which is characterized as a full agonist at the progesterone receptor Purity 99.93%
Catalog No. 50-287-6990 Supplier TARGETMOL CHEMICALS INC Supplier No. T839525MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Nomegestrol acetate is a widely used progestin-like drug,which is characterized as a full agonist at the progesterone receptor Purity 99.93%