Learn More
Medchemexpress LLC Nomegestrol acetate | 58652-20-3 | MFCD00941390 | 99.7% | 370.48 g·mol-1 | C23H30O4 | 1 ML
SDP
Catalog No. 5000240202
Encompass_Preferred
Nomegestrol acetate is a selective progestogen and progesterone receptor agonist used in research as an analytical standard or reagent. Available as a 10 mM solution in DMSO (1 mL) and in solid form, it has confirmed identity and documentation for laboratory use. CAS: 58652-20-3; formula: C23H30O4; molar mass: 370.48 g·mol-1.
  • High purity (99.7%) suitable for analytical work.
  • Pre-diluted 10 mM solution in DMSO for convenience.
  • Also available as a solid for custom preparation.
  • Certificate of analysis and safety data available.
  • Appropriate for progesterone receptor assays and pharmacology studies.
  • Store per supplier instructions to maintain stability.
Catalog No. 50-002-40202 Supplier Medchemexpress LLC Supplier No. HY105634A10MM 1ML
Nomegestrol acetate is a selective progestogen and progesterone receptor agonist used in research as an analytical standard or reagent. Available as a 10 mM solution in DMSO (1 mL) and in solid form, it has confirmed identity and documentation for laboratory use. CAS: 58652-20-3; formula: C23H30O4; molar mass: 370.48 g·mol-1.
  • High purity (99.7%) suitable for analytical work.
  • Pre-diluted 10 mM solution in DMSO for convenience.
  • Also available as a solid for custom preparation.
  • Certificate of analysis and safety data available.
  • Appropriate for progesterone receptor assays and pharmacology studies.
  • Store per supplier instructions to maintain stability.