Learn More
Medchemexpress LLC Nomegestrol acetate | 58652-20-3 | MFCD00941390 | 99.7% | 370.48 g/mol | C23H30O4 | 50 MG
SDP
Catalog No. 5000240185
Encompass_Preferred
Nomegestrol acetate is an orally active, highly selective progestogen and progesterone receptor full agonist supplied for research use. It is provided as a solid for preclinical and in vitro endocrine and receptor pharmacology studies.
  • Orally active, highly selective progestogen.
  • Progesterone receptor full agonist useful for receptor pharmacology.
  • High purity suitable for analytical and preclinical studies.
  • Solid, white to off-white appearance for stable handling and storage.
  • Available in multiple sizes for flexible experimental scale.
  • Defined storage conditions to maintain compound stability.
Catalog No. 50-002-40185 Supplier Medchemexpress LLC Supplier No. HY105634A50MG
Nomegestrol acetate is an orally active, highly selective progestogen and progesterone receptor full agonist supplied for research use. It is provided as a solid for preclinical and in vitro endocrine and receptor pharmacology studies.
  • Orally active, highly selective progestogen.
  • Progesterone receptor full agonist useful for receptor pharmacology.
  • High purity suitable for analytical and preclinical studies.
  • Solid, white to off-white appearance for stable handling and storage.
  • Available in multiple sizes for flexible experimental scale.
  • Defined storage conditions to maintain compound stability.