Learn More
Selleck Chemical LLC Nomegestrol Acetate-E0707-5MG
SDP
Catalog No. 503137046
Encompass_Preferred
Nomegestrol Acetate is a potent highly selective progestogen which is characterized as a full agonist at the progesterone receptor with no or minimal binding to other steroid receptors including the androgen and glucocorticoid receptors
Catalog No. 50-313-7046 Supplier Selleck Chemical LLC Supplier No. E07075MG
Nomegestrol Acetate is a potent highly selective progestogen which is characterized as a full agonist at the progesterone receptor with no or minimal binding to other steroid receptors including the androgen and glucocorticoid receptors