Learn More
Lakeland Industries Nomex Coverall
Supplier:  Lakeland Industries C020135032
Encompass_Preferred
Lakeland Inherently FR 6.0 oz. Nomex coveralls are built tough and comfortable to protect the wearers from flash fires. C020 coveralls are configured with a quick release zipper, heavy duty snaps, pockets, and an elasticized waistband. Available in Navy and Khaki, sizes SM-6X, this coverall is ideal when flash fire hazards are present.
Catalog No. 50-284-0937