Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark™ Professional Nomex Hood

Catalog No. 19168475
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-168-475 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-168-475 Supplier Kimberly-Clark™ Professional Supplier No. 14504

Please call customer service for current pricing. 1-800-766-7000

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kimberly-Clark™ Professional Nomex Hood provides extra protection around the face while wearing a hard hat or welding helmet

  • Provides protection against rain, sleet, snow and cold while wearing a hard hat or welding helmet
  • Made with 100% Nomex
  • Double-layered knotted hood features full head and neck coverage with an extra-wide collar
  • Shoulder-length collar can be tucked securely into a jacket
  • Nomex material is inherently flame resistant, making the hood ideal for industrial environments
  • Can be worn under a hard hat or welding helmet as protection against flame and electrical arcs
  • Retains the flame-resistant protection throughout the life of the product
  • Ideal for cool weather environments
  • Coded for a temperature range of 32 to 45°F (0 to 8°C)

Specifications

Size Universal
Color White
Product Type Arc Flash Hood
For Use With (Application) Protection against flame and electrical arcs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.