Learn More
Aviva Systems Biology Corp NOMO1 Antibody - C-terminal region (ARP54998_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54998_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOMO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NOMO1. Alternate names: Nomo, PM5, NOMO3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61392