Learn More
MSC NON-ACID BTHRM DISINFCTNT
Catalog No. 502819298
Encompass
This non-acid disinfectant bathroom cleaner concentrate is designed for effective cleaning and disinfection in healthcare, educational, and commercial facilities. It is formulated to be diluted and dispensed, providing broad-spectrum efficacy against bacteria, fungi, mildew, and viruses on hard, non-porous surfaces.
  • Concentrated formula for cost-effective use
  • Effective against Pseudomonas aeruginosa, Staphylococcus aureus, Salmonella enterica
  • Controls mold and mildew (Aspergillus niger) and associated odors
  • Fungicidal against Trichophyton mentagrophytes and Candida albicans
  • Virucidal against HIV-1, HBV, HCV, SARS-CoV-2, Human Coronavirus, and Influenza Type A2/Hong Kong Virus
  • EPA registered for disinfection
Catalog No. 50-281-9298 Supplier MSC Supplier No. 14162242
This non-acid disinfectant bathroom cleaner concentrate is designed for effective cleaning and disinfection in healthcare, educational, and commercial facilities. It is formulated to be diluted and dispensed, providing broad-spectrum efficacy against bacteria, fungi, mildew, and viruses on hard, non-porous surfaces.
  • Concentrated formula for cost-effective use
  • Effective against Pseudomonas aeruginosa, Staphylococcus aureus, Salmonella enterica
  • Controls mold and mildew (Aspergillus niger) and associated odors
  • Fungicidal against Trichophyton mentagrophytes and Candida albicans
  • Virucidal against HIV-1, HBV, HCV, SARS-CoV-2, Human Coronavirus, and Influenza Type A2/Hong Kong Virus
  • EPA registered for disinfection