Learn More
SAI Infusion Technologies 22 Gauge, 0.75" Straight Non-Coring Huber Needles - 25/pk, Sterile
SDP
Catalog No. NC0925470
Encompass
Non-coring Huber needles, 22G, 3/4 inch, 25/pack, sterile. These straight non-coring Huber Needles are great for dosing or sampling. Use them with an access port or injection cap. Deflected point allows for greater septum life. 90 Degree Non-Coring Huber Needles and Winged Infusion Sets are available separately. Product is for laboratory animal research only.
Catalog No. NC0925470 Supplier SAI Infusion Technologies Supplier No. H2275
Non-coring Huber needles, 22G, 3/4 inch, 25/pack, sterile. These straight non-coring Huber Needles are great for dosing or sampling. Use them with an access port or injection cap. Deflected point allows for greater septum life. 90 Degree Non-Coring Huber Needles and Winged Infusion Sets are available separately. Product is for laboratory animal research only.