Learn More
Braintree Scientific Non-Coring Huber Needles, Straight, 22g, 0.75"""" (25/pk)
SDP
Catalog No. 501954909
Encompass
Straight Non-Coring Huber Needles - Straight, 22g, 0.75" (25/pk) Great for dosing or sampling. Use with an access port or injection cap Special point allows for greater septum life. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4909 Supplier Braintree Scientific Supplier No. H2275
Straight Non-Coring Huber Needles - Straight, 22g, 0.75" (25/pk) Great for dosing or sampling. Use with an access port or injection cap Special point allows for greater septum life. **Sold for research purposes only and not for human use**