Learn More
MSC Dow Corning 99180561 10.1 oz Tube Off-White RTV Silicone Joint Sealant -67 to 350°F Operating Temp, 30 min Tack Free Dry Time, 36 hr Full Cure Time, Series 748
Catalog No. 501305379
Encompass
Dow Corning 99180561 10.1 oz Tube Off-White RTV Silicone Joint Sealant -67 to 350°F Operating Temp, 30 min Tack Free Dry Time, 36 hr Full Cure Time, Series 748
Catalog No. 50-130-5379 Supplier MSC Supplier No. 00232041
Dow Corning 99180561 10.1 oz Tube Off-White RTV Silicone Joint Sealant -67 to 350°F Operating Temp, 30 min Tack Free Dry Time, 36 hr Full Cure Time, Series 748