Learn More
MSC Dow Corning 2455846 3 oz Tube Off-White RTV Silicone Joint Sealant -67 to 350°F Operating Temp, 30 min Tack Free Dry Time, 36 hr Full Cure Time, Series 748
Catalog No. 501149234
Encompass
Dow Corning 2455846 3 oz Tube Off-White RTV Silicone Joint Sealant -67 to 350°F Operating Temp, 30 min Tack Free Dry Time, 36 hr Full Cure Time, Series 748
Catalog No. 50-114-9234 Supplier MSC Supplier No. 00232025
Dow Corning 2455846 3 oz Tube Off-White RTV Silicone Joint Sealant -67 to 350°F Operating Temp, 30 min Tack Free Dry Time, 36 hr Full Cure Time, Series 748