Learn More
Med Vet International Non-Cuffed Endotracheal Tube, J0149U, 3.0mm, Each
Catalog No. 5000583692
Encompass_Preferred
Non-Cuffed Endotracheal Tube. Softens at body temperature, reducing postop- erative irritation. Blue-tinted PVC tube complete with connector. Available in 0.5mm increments for 3.5mm to 13mm.Tube. 3.0mm, Length 16cm
Catalog No. 50-005-83692 Supplier Med Vet International Supplier No. JOR149U
Non-Cuffed Endotracheal Tube. Softens at body temperature, reducing postop- erative irritation. Blue-tinted PVC tube complete with connector. Available in 0.5mm increments for 3.5mm to 13mm.Tube. 3.0mm, Length 16cm