Learn More
Invent Biotechnologies Inc Minute™ Non-Denatured Protein Solubilization Reagent (10 ml)
Catalog No. NC1628013
Encompass
This item has a minimum qty of 4 per supplier requirements.
This reagent is formulated for dissolving protein pellets using protein kits from Invent Biotechnologies (Cat# Nos. SM-005, SM-005-P, CP-011, NE-013, YM-017, MM-018 and AF-023) and other companies. Solubilized proteins are non-denatured and can be used for ELISA, immunoprecipitation/Co-IP, enzymatic activity determination and other applications.
Catalog No. NC1628013 Supplier Invent Biotechnologies Inc Supplier No. WA010
This item has a minimum qty of 4 per supplier requirements.
This reagent is formulated for dissolving protein pellets using protein kits from Invent Biotechnologies (Cat# Nos. SM-005, SM-005-P, CP-011, NE-013, YM-017, MM-018 and AF-023) and other companies. Solubilized proteins are non-denatured and can be used for ELISA, immunoprecipitation/Co-IP, enzymatic activity determination and other applications.