Learn More
Biotium Dry Milk Powder, 4 x 25 g
Catalog No. 501964394
Encompass_Preferred
Nonfat dry milk is commonly added to blocking buffers minimize background noise in immunodetection procedures such as Western blotting and immunostaining. Milk can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or used for blocking and dilution of antibodies during immunostaining procedures.
Catalog No. 50-196-4394 Supplier Biotium Supplier No. 22012
Nonfat dry milk is commonly added to blocking buffers minimize background noise in immunodetection procedures such as Western blotting and immunostaining. Milk can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or used for blocking and dilution of antibodies during immunostaining procedures.