Learn More
MSC Detco Glass Cleaner, Invisi-Blu, Concentrated, 9:1 Dilution, 32 oz
Catalog No. 502816153
Encompass
Invisi-Blu is a concentrated glass cleaner designed for efficient and effective cleaning of glass surfaces. Supplied in a 32 oz bottle, this water-based formula offers a 9:1 dilution ratio.
  • Biodegradable for environmentally conscious cleaning
  • Unscented formula for versatile application
  • Water-based for safe use on glass surfaces
  • Concentrated for cost-effective use
  • Provided in a convenient bottle container
Catalog No. 50-281-6153 Supplier MSC Supplier No. 33687476
Invisi-Blu is a concentrated glass cleaner designed for efficient and effective cleaning of glass surfaces. Supplied in a 32 oz bottle, this water-based formula offers a 9:1 dilution ratio.
  • Biodegradable for environmentally conscious cleaning
  • Unscented formula for versatile application
  • Water-based for safe use on glass surfaces
  • Concentrated for cost-effective use
  • Provided in a convenient bottle container