Learn More
Creative Biomart Ancestim, Recombinant human stem cell factor
Supplier:  Creative Biomart THP0047
Encompass_Preferred
The product is a non-glycosylated recombinant methionyl human stem cell factor. It is a 166 amino acid protein produced by E. coli with an amino acid sequence that is identical to the natural sequence predicted from human DNA sequence analysis, except for the addition of an N-terminal methionine.
Catalog No. 50-208-4098