Learn More
Grainger Brady® Non-Hazardous Waste Labels, B-235 Paper, 3.5 in x 10 in, 100/Pk.
Catalog No. NC9861363
Encompass_Preferred
These non-hazardous waste labels are designed for clear facility and safety identification. Featuring semi-gloss B-235 coated paper, these rectangular labels use a pressure-sensitive permanent acrylic adhesive to adhere to most general surfaces while maintaining the ability to be removed cleanly without leaving residue.
  • Features white text on a green background reading Non-Hazardous Waste
  • Constructed from semi-gloss B-235 paper material
  • Provides service temperatures from -40 °F to 150 °F
  • Measures 88.90 mm (3.5 in.) wide by 254.00 mm (10 in.) high
  • Designed for indoor use
  • Includes 100 labels per pack
Catalog No. NC9861363 Supplier Grainger Supplier No. 8AAR3
These non-hazardous waste labels are designed for clear facility and safety identification. Featuring semi-gloss B-235 coated paper, these rectangular labels use a pressure-sensitive permanent acrylic adhesive to adhere to most general surfaces while maintaining the ability to be removed cleanly without leaving residue.
  • Features white text on a green background reading Non-Hazardous Waste
  • Constructed from semi-gloss B-235 paper material
  • Provides service temperatures from -40 °F to 150 °F
  • Measures 88.90 mm (3.5 in.) wide by 254.00 mm (10 in.) high
  • Designed for indoor use
  • Includes 100 labels per pack