Learn More
Perkin Elmer US LLC Non HF-Resistant Internal Standard Kit
Catalog No. 509049658
Encompass_Preferred
All connections to the mixing chamber/combiner use EzyFit connectors. A sampling probe for the reagent addition bottle is also included so that it remains well anchored in the bottle. This kit is completely modular so that damaged or worn components can easily be replaced. This kit includes: EzyFit Glass Mixing T-piece, Qty. 1 (N0777461) Internal Standard Sample Probe 0.25 mm I.D., Qty. 1 (N0777465) EzyFit Sample Tube 0.5 mm I.D., Qty. 1 (N0777463) EzyFit Sample Tube – Twin, Qty. 1 (N0777462) EzyFit Internal Standard Tube 0.25 mm I.D., Qty. 1 (N0777464) Vitex Gripper Paper, Qty. 6 (N0777848) Orange/Blue Flared End PVC Tubing 0.25 mm I.D., Qty. 12 (N0773112) UniFit-EzyFit Sample Tube, Qty. 1 (N0777061)
Catalog No. 50-904-9658 Supplier Perkin Elmer US LLC Supplier No. N0774068
All connections to the mixing chamber/combiner use EzyFit connectors. A sampling probe for the reagent addition bottle is also included so that it remains well anchored in the bottle. This kit is completely modular so that damaged or worn components can easily be replaced. This kit includes: EzyFit Glass Mixing T-piece, Qty. 1 (N0777461) Internal Standard Sample Probe 0.25 mm I.D., Qty. 1 (N0777465) EzyFit Sample Tube 0.5 mm I.D., Qty. 1 (N0777463) EzyFit Sample Tube – Twin, Qty. 1 (N0777462) EzyFit Internal Standard Tube 0.25 mm I.D., Qty. 1 (N0777464) Vitex Gripper Paper, Qty. 6 (N0777848) Orange/Blue Flared End PVC Tubing 0.25 mm I.D., Qty. 12 (N0773112) UniFit-EzyFit Sample Tube, Qty. 1 (N0777061)