Learn More
Non Ionic Surfactants in H2O, PT Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111004824 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
Catalog No. Quantity
11-100-4824 20 mL
1 options
Catalog No. 11-100-4824 Supplier MilliporeSigma Supelco Supplier No. PE119720ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This PT sample is as a 20mL concentrate. All constituents are present in soluble form. The solvent for this sample is water.Sample PreparationPartially fill a 1000-mL volumetric flask with reagent-grade water (near 20C). Open ampule PEI-088 and transfer 10.0 mL of concentrate to the volumetric flask. Fill the volumetric flask to volume with reagent-grade water and mix well. The sample is now ready for immediate analysis
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 20 mL
Concentration or Composition (by Analyte or Components) Surfactant - Nonionic 0.2-1 mg/L