Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Non-Magnetic Fixed Gurney

Catalog No. 22505092
Change view
Click to view available options
Capacity (English):
600 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22-505-092 600 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-092 Supplier Marketlab Supplier No. 240732
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-magnetic, bariatric stretcher with 600 lb. weight capacity and drop-down side rails.

  • Includes 11 in. drop-down side rails for easy accessibility and patient safety.

Specifications

Capacity (English) 600 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.