Learn More
Stoelting™ Non-Puncture Cat/Monkey Ear Bars
SDP
Catalog No. 10000144 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Non-rupture
Catalog No. Model
10-000-144 Non-rupture
1 options
Catalog No. 10-000-144 Supplier Stoelting™ Supplier No. 51614
Add to Cart
Add to Cart
Stoelting™ Non-Puncture Cat/Monkey Ear Bars have a 45° taper and do not puncture the tympanic membrane.
Compatible with all Lab Standard Series Rat Stereotaxic instruments
Specifications
For Use With (Application) Neuroscience
Size 45° taper
Animal Cat, Monkey
Model Non-rupture