Learn More
SOMNI Scientific™ Non-Rebreathing Small Animal Anesthesia System
SDP
Catalog No. 04777109 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-109 Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-109 Supplier SOMNI Scientific™ Supplier No. RM17004
Add to Cart
Add to Cart
Made of high-quality materials, supports an O2 concentrator and has a small footprint
  • High quality limited maintenance design
  • H-frame with shelf provides support and stability and the shelf adds vital workspace
  • Integrated common gas outlet for breathing circuit; no adapters needed
  • 3 in. wheels for maneuverability
  • Supports oxygen concentrator
  • Small footprint, easy assembly
  • Powder coated and anodized frame for easy cleaning
Specifications
For Use With (Application) Animal Inhalant Anesthesia
For Use With (Equipment) VS-6002, VS-6001,VS-6004, VS-6006, VS-6005, VS-6008, OC-21003, RE-20101, WG-15003, DS-14005
Material Powder-coated and Anodized Aluminum
Length (Metric) 53.34 cm
Width (Metric) 53.34 cm
Includes Non-rebreathing circuit and bag
For Veterinary Use Only