Learn More
Pyramex™ Non-Reflective Hi-Vis Safety Vests
Catalog No. 19555444 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-444 Hi-vis Orange
19-555-442 Hi-vis Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-555-444 Supplier Pyramex™ Supplier No. RV120NS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Non-Reflective Hi-Vis Safety Vests do not meet ASNI/CSA safety standards
  • Front: 5 in. Hook & Loop closure
  • Sides: 0.75 x 7 in. elastic bands
Specifications
Closure Type Hook and Loop (5 in.)
Color Hi-vis Orange
Size One Size
Material Polyester Mesh
No. per Case 12/Cs.