Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Non-Reinforced Surgical Gowns

Catalog No. 19162301 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Small/Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-301 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-162-301 Supplier Cardinal Health™ Supplier No. 9505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AAMI Level 3 surgical gowns are constructed with SMS material and offer outstanding barrier protection.

  • AAMI Level 3
  • Standard
  • Latex-free
  • Non-reinforced

Additional Information:

  • Sterility: Sterile
  • Latex content: Latex-Free
  • Feature/Property: Set-In Sleeve, AAMI Level 3, Standard, Non-reinforced, Sterile

Specifications

For Use With (Application) Surgical
Product Type Surgical Gown
Gender Unisex
No. of Pockets None
Size Small/Medium
Material SMS
Color Royal Blue
Closure Type Back Ties
Disposable Single-use
Sterility Sterile
Cuff Style Knit
Latex Free Yes
Sleeve Style Set-in Sleeve
Certifications/Compliance AAMI Level 3, 16 CFR Part 1610
Autoclavable Not Autoclavable
No. per Pack 20/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.