Learn More
MSC Mitutoyo Non-Rotating Disc Micrometer, 2 in. to 3 in. Range, 0.00005 in. Resolution, IP65, 1 ea.
Catalog No. 5000513039
Encompass
This non-rotating disc micrometer is designed for precise measurements of gear teeth and paper thickness. It features an IP65-rated dust and water protection level, ensuring reliable operation in harsh workshop environments. The device offers high-accuracy measurement with a wide range and clear LCD display for easy reading.
  • Carbide tipped measuring faces for enhanced durability
  • Graduation of .00005 in. (0.001 mm)
  • Measurement range of 2 in. to 3 in.
  • Convenient inch/mm conversion capabilities
  • Includes auto power on/off and data output functionality
  • Long battery life of approximately 2.4 years under normal use
  • Features zero/ABS, origin-set, and hold functions
Catalog No. 50-005-13039 Supplier MSC Supplier No. 74352949
This non-rotating disc micrometer is designed for precise measurements of gear teeth and paper thickness. It features an IP65-rated dust and water protection level, ensuring reliable operation in harsh workshop environments. The device offers high-accuracy measurement with a wide range and clear LCD display for easy reading.
  • Carbide tipped measuring faces for enhanced durability
  • Graduation of .00005 in. (0.001 mm)
  • Measurement range of 2 in. to 3 in.
  • Convenient inch/mm conversion capabilities
  • Includes auto power on/off and data output functionality
  • Long battery life of approximately 2.4 years under normal use
  • Features zero/ABS, origin-set, and hold functions