Learn More
World Precision Instrument Non-Rupture Ear Bars and Serrated Cuffs for Mouse Stereotaxic
SDP
Catalog No. 502539148
Encompass
Using model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The Mouse/Neonatal Rat Adaptor is designed for use with the traditional 'U' frame Stereotaxic instrument. The adaptor employs light, Delrin Ear Bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups.
Catalog No. 50-253-9148 Supplier World Precision Instrument Supplier No. 505268
Using model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The Mouse/Neonatal Rat Adaptor is designed for use with the traditional 'U' frame Stereotaxic instrument. The adaptor employs light, Delrin Ear Bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups.