Learn More
OHAUS Non-Skid Tray for Enviro-Genie
Catalog No. 01804358 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-358 Shaker Attachment
1 options
Catalog No. 01-804-358 Supplier OHAUS Supplier No. 83041425
Only null left
Add to Cart
Add to Cart
The Non-Skid Tray for the Enviro-Genie and the Incubator-Genie attaches to the Magnetic Platform (Part No SI-1320) to rock open containers.
  • The Non-Skid Tray measures 203 x 305 mm (8 x 12 in.) and is included with the Enviro-Genie and Incubator-Genie
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment
Dimensions (L x W) 203 x 305 mm