Learn More
Bio Plas Non-skirted Low Profile Universal PCR Plate
Catalog No. 22038957
Change view
Click to view available options
Color:
Clear
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-957 White
22-038-956 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-038-957 Supplier Bio Plas Supplier No. 9409
Add to Cart
Add to Cart
Low profile PCR plate without skirt.
  • 0.1 mL
  • Universal PCR plate
  • Non-Skirted
  • Low Profile
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Not Autoclavable
Color White
Disposable Single-use
Product Type PCR Plate
Material Polycarbonate
Volume (Metric) Well 1.0 mL
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
RNase Free Yes
Fit for Purpose PCR Ready