Learn More
Electron Microscopy Sciences Non Slip Forceps with Serrated Straight Tips, 175 mm, 0.75 mm Tip width
SDP
Catalog No. 5030232
Encompass_Preferred
This outstanding design fits nicely into all hands with perfect functionality. The forceps offer a light touch and are non-slip with high precision finish tips.
OAL: 175 mm
Tip width: 0.75 mm
Catalog No. 50-302-32 Supplier Electron Microscopy Sciences Supplier No. 78301S
This outstanding design fits nicely into all hands with perfect functionality. The forceps offer a light touch and are non-slip with high precision finish tips.
OAL: 175 mm
Tip width: 0.75 mm