Learn More
OHAUS Non-Slip Mat for Low Speed
Catalog No. 01804366 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-366 Shaker Attachment
1 options
Catalog No. 01-804-366 Supplier OHAUS Supplier No. 83041348
Gray silicone mat is an alternative to flask clamps or roller bars for securing vessels to shaker platform of Orbital-Genie Shaker for low speed.
  • Alternative to flask clamps or roller bars.
  • For securing vessels to shaker platform of Orbital-Genie Shaker for low speed.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment