Learn More
Electron Microscopy Sciences Kimball Physics Non-Standard Selective LaB6 Cathode
SDP
Catalog No. 5026874
Encompass
LaB6 Cathodes are high performance, resistively heated, thermionic electron source for use in SEM, TEM, electronlithography systems, and x-ray sources. They offer an extended life with thousands of hours in clean vacuum, exceptional thermal, chemical, and electrical stability, high brightness/low energy spread with oriented single crystal made from the best quality/high purity material. 60 deg angle w/non-standard microflat
Catalog No. 50-268-74 Supplier Electron Microscopy Sciences Supplier No. 8108060
LaB6 Cathodes are high performance, resistively heated, thermionic electron source for use in SEM, TEM, electronlithography systems, and x-ray sources. They offer an extended life with thousands of hours in clean vacuum, exceptional thermal, chemical, and electrical stability, high brightness/low energy spread with oriented single crystal made from the best quality/high purity material. 60 deg angle w/non-standard microflat