Learn More
Tisch Scientific Nylon Syringe Filters, 5 um, 25mm, Luer-Lok/Luer Slip, Nonsterile, 100 per pack, SF15156
SDP
Supplier:  Tisch Scientific SF15156
Encompass
Tisch nylon syringe filters provide a universal application for analytical procedures. Naturally hydrophilic, high protein binding and high dirt loading capability, and good chemical resistance make them ideal for aqueous (non-acidic) or organic sample preparations, HPLC, GC or dissolution sample analysis.
Catalog No. NC1770001