Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Non-Sterile BucketLiner
BIOPHARMASDP

Catalog No. 19116741 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Dimensions:
31 x 19 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions
19-116-741 -
19-065-150 31 x 19 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-116-741 Supplier Micronova™ Supplier No. BKL2424
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Micronova™ Non-Sterile BucketLiner creates an impermeable barrier between strong disinfectants and buckets or between product and container.

  • Laminate is 75% LDPE, 25% Nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts or raw material transfer
  • 3 mil thickness
  • Flat packed, individually double-bagged or bulk packed
  • Holds up to autoclave with liquid contents
  • Non-Sterile liners are bulk packed 50 per bag, 200 Liners per case

Specifications

Material LDPE, Nylon
Autoclavable Not Autoclavable
Color Clear
For Use With (Application) Product Transport or Liquid Containment
For Use With (Equipment) B-7 series Bucket or Similar Sized tote
Length (Metric) 60 cm
Quantity 24/Pk.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.