Learn More
Cincinnati Surgical Non-Sterile Carbon Blades
SDP
Catalog No. 19200261
Change view
Click to view available options
Size:
22
70
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-200-261 22
19-200-256 70
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-200-261 Supplier Cincinnati Surgical Supplier No. 0222
Add to Cart
Add to Cart
This item is not returnable. View return policy
Packaged with VCI Paper to protect blade from corrosion
  • Made from high-grade British surgical carbon steel
  • Narrow grinding angle resulting in increased initial keenness
  • Medium surface finish
  • 200% visual inspection
  • Strong foil package
Specifications
Blade Material British Carbon Steel
Size 22
Sterility Non-sterile