Learn More
Andwin Scientific NON-STERILE PIPETTE 100/PK
SDP
Catalog No. 50930631
Encompass_Preferred
With a length of 30.0 centimeters, the extra long transfer pipettes are perfect for subculturing, harvesting, sampling 15, 50, 100 ml tubes and flask, removing supernatant and media from cell cultures, washing cells, post centrifugation to remove cell layers, layering in columns or transfering in columns, loading gels, field testing, blood culture bottle, and several other purposes. This product is sold in a bulk quantity of 100 non-sterile pipettes.
Catalog No. 50-930-631 Supplier Andwin Scientific Supplier No. 7096827
With a length of 30.0 centimeters, the extra long transfer pipettes are perfect for subculturing, harvesting, sampling 15, 50, 100 ml tubes and flask, removing supernatant and media from cell cultures, washing cells, post centrifugation to remove cell layers, layering in columns or transfering in columns, loading gels, field testing, blood culture bottle, and several other purposes. This product is sold in a bulk quantity of 100 non-sterile pipettes.