Learn More
Andwin Scientific NON-STERILE PIPETTE 400/PK
SDP
Catalog No. 50930594
Encompass_Preferred
The disposable transfer pipets are ideal for general laboratory use. This product can be utilized for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The pipets are sold in a bulk quantity of 500 pieces.
Catalog No. 50-930-594 Supplier Andwin Scientific Supplier No. 709602
The disposable transfer pipets are ideal for general laboratory use. This product can be utilized for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The pipets are sold in a bulk quantity of 500 pieces.