Learn More
Andwin Scientific NON-STERILE PIPETTE 500/PK
SDP
Supplier:  Andwin Scientific 7096210
Encompass_Preferred
The graduated transfer pipets are specifically made to be utilized for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, and several other applications. This product has a measured length of 15.2 centimeters and a stem diameter of 7.1 millimeters. The non-sterile pipets are packed in a bulk of 500 pieces.
Catalog No. 50-930-604