Learn More
KDEB MANUFACTURING 96 Deep Well KDeb Microplate, 2 ml Full Capacity "U" Wells, 8 x 12 Grid Configuration, Distinct Alpha-Numeric Designation, For Use in Automation Processes, Overall Well Plate Dimensions: 128 x 86 x 42mm, Polypropylene
Catalog No. NC1582499
Encompass_Preferred
The KDeb Plate features the same footprint as all standard 96 well microtiter plates and is compatible with commonly used 8 and 12 channel pipettes, compatible for liquid nitrogen storage, and a standard tool in analytical research and diagnostic testing laboratories. The alpha-numeric designation makes the KDeb Plate ideal for storage or archiving of specimens. The KDeb Plate is useful for DNA sequencing, ELISA and procedures requiring a mother plate or block. Other common uses include culture retrieval of bacteria and cell tissue in clinical research applications. Regularly used in automated laboratory processes and for pharmaceutical R&D studies. The non-sterile KDeb Plate, formerly U.S. Riplate, meets the requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.
Catalog No. NC1582499 Supplier KDEB MANUFACTURING Supplier No. 850356BX
The KDeb Plate features the same footprint as all standard 96 well microtiter plates and is compatible with commonly used 8 and 12 channel pipettes, compatible for liquid nitrogen storage, and a standard tool in analytical research and diagnostic testing laboratories. The alpha-numeric designation makes the KDeb Plate ideal for storage or archiving of specimens. The KDeb Plate is useful for DNA sequencing, ELISA and procedures requiring a mother plate or block. Other common uses include culture retrieval of bacteria and cell tissue in clinical research applications. Regularly used in automated laboratory processes and for pharmaceutical R&D studies. The non-sterile KDeb Plate, formerly U.S. Riplate, meets the requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.