Learn More
StatLab Non-sterile Screw Top Urine Specimen Containers
SDP
Catalog No. 21402138
Change view
Click to view available options
Volume (English):
4 oz.
Catalog No. Volume (English)
21-402-138 4 oz.
1 options
Catalog No. 21-402-138 Supplier StatLab Supplier No. CT5019
Add to Cart
Edge
Add to Cart
Urine specimen containers, designed to reduce leakage with non-sterile feature
  • Our StatClick feature is now available in a urine container
  • An audible click indicates that the lid is securely fastened to container
  • 4 oz, 500 per case
Specifications
Product Type Urinalysis Specimen Container
Disposable Single-use
Graduated Yes
Material Polypropylene
Sterility Non-sterile
Volume (English) 4 oz.