Learn More
Cincinnati Surgical Non-sterile Size 60 Surgical Blade
SDP
Catalog No. 14100250
Change view
Click to view available options
Size:
# 60
Catalog No. Size
14-100-250 # 60
1 options
This item is not returnable. View return policy
Catalog No. 14-100-250 Supplier Cincinnati Surgical Supplier No. 0260CS
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Size 60 blades are the blade of choice within of pathology and histopathology
  • Non-sterile
  • Individually foil wrapped
  • 100 blades/Pk.
  • Fits a size 8 handle
Specifications
Product Type Surgical Blade
Size # 60
For Use With (Equipment) Fits # 8 Handle
Disposable Single-use
Sterility Non-sterile
Quantity 10/Pk.