Learn More
Stoelting™ Non-Sterile Sponge Gauze
SDP
Catalog No. p-7163791 Shop All Stoelting Co Products
Change view
Click to view available options
Dimensions (L x W):
2 x 2 in.
3 x 3 in.
4 x 4 in.
Includes:
Case of 2000
Case of 4000
Case of 8000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Includes
10-000-688 2 x 2 in. Case of 8000
10-000-689 3 x 3 in. Case of 4000
10-000-690 4 x 4 in. Case of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-000-688 Supplier Stoelting™ Supplier No. 50971
Only null left
Add to Cart
Add to Cart
Stoelting™ Non-Sterile Sponge Gauze potects and cushions wounds
  • Unique rayon and polyester nonwoven fabric
  • Edges are folded inward to eliminate linting and fraying
  • General-Use sponges provide strength, absorbency and scrubbing texture
  • Promotes quick drainage and full utilization of entire sponge
  • Peel-open package ensures integrity and aseptic preparation
  • 100% cotton
Specifications
For Use With (Application) Neuroscience
Dimensions (L x W) 2 x 2 in.
Model Non-woven sponge
Includes Case of 8000