Learn More
Fine Science Tools Silk Suture Thread, Braided Silk, 3, 0.65 mm, 23 m
SDP
Catalog No. NC9710956
Encompass
Product: Suture Thread - Braided Silk/Size 3/ Size: 3/ Suture Diameter: 0.65mm/ Suture Length: 22.5m/ Suture Material: Braided Silk/ Sterile: No/ Unit: Each
Catalog No. NC9710956 Supplier Fine Science Tools Supplier No. 1802003
Product: Suture Thread - Braided Silk/Size 3/ Size: 3/ Suture Diameter: 0.65mm/ Suture Length: 22.5m/ Suture Material: Braided Silk/ Sterile: No/ Unit: Each