Learn More
Revvity Health Sciences Inc P235 Non-Sterile Tips, Clear, 5-235L
Catalog No. NC1304907
Encompass_Preferred
Robotic Certified Tips Precision Tips for Sample Preparation Liquid handling for lab automation requires more than just generic pipette tips. We offer Robotic Certified Tips for 96- or 384- channel pipettors. Made from 100% virgin polypropylene, our Robotic Certified Tips are inspected with strict tolerances for straightness and uniformity. Gold Standard Quality Control We offer a portfolio of Robotic Certified Tips specially engineered with the highest degree of uniformity thus providing unparalleled accuracy in processing high density microplates for high throughput applications. Made from medical grade 100% virgin polypropylene and manufactured in a clean room environment to eliminate contamination, these Robotic Certified Tips are inspected with strict tolerances for straightness, uniformity and performance. To ensure that our tips meet the most exacting requirements for laboratory automation, each tip goes through a rigorous inspection process before it is certified lab ready. Ea
Catalog No. NC1304907 Supplier Revvity Health Sciences Inc Supplier No. 6001295
Robotic Certified Tips Precision Tips for Sample Preparation Liquid handling for lab automation requires more than just generic pipette tips. We offer Robotic Certified Tips for 96- or 384- channel pipettors. Made from 100% virgin polypropylene, our Robotic Certified Tips are inspected with strict tolerances for straightness and uniformity. Gold Standard Quality Control We offer a portfolio of Robotic Certified Tips specially engineered with the highest degree of uniformity thus providing unparalleled accuracy in processing high density microplates for high throughput applications. Made from medical grade 100% virgin polypropylene and manufactured in a clean room environment to eliminate contamination, these Robotic Certified Tips are inspected with strict tolerances for straightness, uniformity and performance. To ensure that our tips meet the most exacting requirements for laboratory automation, each tip goes through a rigorous inspection process before it is certified lab ready. Ea