Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Unity Lab Services™ Non-USP/EP UV-Vis Qualified Standards Rental Fee

Catalog No. 701061061 Shop All Thermo Scientific Products
Change view
Click to view available options
Duration:
1 Day
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Duration
70-106-1061 1 Day
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 70-106-1061 Supplier Unity Lab Services™ Supplier No. 701061061
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rent standard to perform PV tests in the UV and visible region

  • Contains standards used to perform PV tests in the UV and visible region
  • Includes: Potassium dichromate solutions (0, 60, and 140 mg/L) for UV absorbance photometric accuracy to 2A, holmium oxide glass filter for wavelength accuracy determination from 241 to 641 nm, didymium glass filter for wavelength accuracy determination from 500 to 900 nm, neutral density filters (4) calibated at 440 nm for photometric repeatability tests between 0.3 and 1.5 A, stray radiant energy filter for stray light detection at 220 nm
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Evolution One, Evolution One Plus, Evolution Pro
Duration 1 Day
Product Type Installation Service
Product Line Thermo Scientific
Service Offering USP Standards Rental
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.