Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Non-USP/EP UV-Vis Qualified Standards Set

Catalog No. 699141800 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
69-914-1800 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 69-914-1800 Supplier Thermo Scientific™ Supplier No. 699141800
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains standards used to perform PV tests in the UV and visible region

  • Contains standards used to perform PV tests in the UV and visible region
  • Includes: Potassium dichromate solutions (0, 60, and 140 mg/L) for UV absorbance photometric accuracy to 2A, holmium oxide glass filter for wavelength accuracy determination from 241 to 641 nm, didymium glass filter for wavelength accuracy determination from 500 to 900 nm, neutral density filters (4) calibated at 440 nm for photometric repeatability tests between 0.3 and 1.5 A, stray radiant energy filter for stray light detection at 220 nm
TRUSTED_SUSTAINABILITY

Specifications

Technique UV/Vis, Spectrometry, LC/MS, HPLC
Color Black
For Use With (Equipment) LCMS, GCMS, HPLC, GC and UV/Vis Spectrotometer
Solution Type Calibration Standard
Type UV Internal Standard
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.