Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Non-Vacuum Glass Desiccator
GREENER_CHOICESDP

Catalog No. 31502264 Shop All Kemtech Products
Change view
Click to view available options
Volume (Metric):
1.5 L
3 L
4.5 L
7 L
13 L
25 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
31-502-264 1.5 L
31-502-265 3 L
31-502-266 4.5 L
31-502-267 7 L
31-502-268 13 L
31-502-269 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-502-264 Supplier Synthware™ Supplier No. B135115
Add to Cart
Add to Cart

Clear, borosilicate Non-Vacuum Glass Desiccator includes a lid and ceramic plate.

  • Non-vacuum desiccator.
  • Made of borosilicate glass.
  • Includes a lid and ceramic plate.

Specifications

Color Clear
Material Borosilicate Glass (3.3)
Product Type Nonvacuum Desiccator
Includes Ceramic Plate, Lid
Volume (English) 0.4 gal.
Volume (Metric) 1.5 L
Height (English) 4.3 in.
Height (Metric) 110 mm
Diameter (Metric) Outer 150 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.