Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Non-Vented Short Brim Hard Hat

Catalog No. 19185000
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-185-000 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-185-000 Supplier Honeywell™ North™ Supplier No. NSB10001

missing translation for 'validMainframeMsg'

Cost-effective certified head protection solution offers enhanced performance, all-day comfort and durability

  • High-density polyethylene (HDPE)
  • Four-point ratchet suspension
  • Dual-locking mechanism
  • Men's
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI (ANSI Z89.1 – 2014) standard, CE/EN (EN 397:2012 +AI:2012, EN 50365:2002 (non-vented versions) standard, CSA (CSA Z94.1-15) standard, NOM standard
Color White
For Use With (Equipment) CP5005 Plastic Faceshield Bracket, 3796VS1 Helmet Adapter
For Use With (Application) Construction, Mining, Manufacturing, Oil and gas, Ship building, Traffic safety, Utilities
Material HDPE
Product Type Type 1
Size Universal
Ventilated Not Ventilated
Head Style Short Brim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.